2-Chlor-4-(trifluormethyl)benzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-4-(trifluoromethyl)benzylalcohol

AS32409G1 AS32409G5
ACTIAS32409G1EA 0 CHF
ACTIAS32409G1 ACTIAS32409G5
2-Chlor-4-(trifluormethyl)benzylalkohol
2-Chlor-4-(trifluormethyl)benzylalkohol
Formel: C₈H₆ClF₃O
MW: 210,58 g/mol
MDL Number: MFCD04972757
CAS-Nummer: 56456-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR