2-(Methylsulfonyl)benzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS20700G25 AS20700G5
ACTIAS20700G25EA 0 CHF
ACTIAS20700G25 ACTIAS20700G5
2-(Methylsulfonyl)benzoesäure
2-(Methylsulphonyl)benzoic acid
Formel: C₈H₈O₄S
MW: 200.21 g/mol
MDL Number: MFCD00216492
CAS-Nummer: 33963-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR