2-Chlor-4-(methylsulfonyl)benzoesäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS22437G100EA 64 CHF
ACTIAS22437G100
2-Chlor-4-(methylsulfonyl)benzoesäure
2-Chloro-4-(methylsulphonyl)benzoic acid
Formel: C₈H₇ClO₄S
MW: 234,66 g/mol
MDL Number: MFCD00216496
CAS-Nummer: 53250-83-2
EINECS: 406-520-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR