2-Chlor-4-(methylsulfonyl)benzoesäure 95%
Lieferant: Thermo Scientific
|
Gefahr
|
ACRO425010010EA
48.1
CHF
ACRO425010010
2-Chlor-4-(methylsulfonyl)benzoesäure 95%
2-Chloro-4-(methylsulphonyl)benzoic acid
Formel:
C₈H₇ClO₄S MW: 234,66 g/mol Melting Pt: 192…193 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00216496 CAS-Nummer: 53250-83-2 EINECS: 406-520-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow to light pink Powder |
Infrared spectrum | Conforms |
Melting point | 191 to 196 °C |
Loss on drying | ≤5 % |
Titration with NaOH | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...