4-Brom-2-(methylsulfonyl)benzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS22588G1 AS22588G5
ACTIAS22588G1EA 0 CHF
ACTIAS22588G1 ACTIAS22588G5
4-Brom-2-(methylsulfonyl)benzoesäure
4-Bromo-2-(methylsulphonyl)benzoic acid
Formel: C₈H₇BrO₄S
MDL Number: MFCD04037926
CAS-Nummer: 142994-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR