3-Brom-4-(methylsulfonyl)benzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS22475G1
AS22475G25
AS22475G5
ACTIAS22475G1EA
0
CHF
ACTIAS22475G1
ACTIAS22475G25
ACTIAS22475G5
3-Brom-4-(methylsulfonyl)benzoesäure
3-Bromo-4-(methylsulphonyl)benzoic acid
Formel:
C₈H₇BrO₄S MW: 279.1100 g/mol |
MDL Number:
MFCD13191607 CAS-Nummer: 39058-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...