4-Pyrid-4-ylbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Pyridin-4-yl-benzoic acid

AS30234G1 AS30234G5
ACTIAS30234G1EA 0 CHF
ACTIAS30234G1 ACTIAS30234G5
4-Pyrid-4-ylbenzoesäure
4-Pyrid-4-ylbenzoesäure
Formel: C₁₂H₉NO₂
MDL Number: MFCD04114574
CAS-Nummer: 4385-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR