4-Pyrid-3-ylbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Pyridin-3-yl-benzoic acid

AS30233G1 AS30233G5
ACTIAS30233G1EA 0 CHF
ACTIAS30233G1 ACTIAS30233G5
4-Pyrid-3-ylbenzoesäure
4-Pyrid-3-ylbenzoesäure
Formel: C₁₂H₉NO₂
MDL Number: MFCD03426513
CAS-Nummer: 4385-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR