4-Pyrid-2-ylbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(2-Pyridyl)benzoic acid, 4-(Pyridin-2-yl)benzoic acid

AS30232G1 AS30232G5
ACTIAS30232G1EA 0 CHF
ACTIAS30232G1 ACTIAS30232G5
4-Pyrid-2-ylbenzoesäure
4-Pyrid-2-ylbenzoesäure
Formel: C₁₂H₉NO₂
MDL Number: MFCD01830093
CAS-Nummer: 4385-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR