1-Ethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(1-Ethyl-1H-pyrazol-4yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Ethyl-1H-pyrazole-4-boronic acid Pinacol ester, 1-Ethyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

AS22364G1 AS22364G25 AS22364G5
ACTIAS22364G1EA 0 CHF
ACTIAS22364G1 ACTIAS22364G25 ACTIAS22364G5
1-Ethyl-1H-pyrazol-4-boronsäurepinakolester
1-Ethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
MDL Number: MFCD09864188
CAS-Nummer: 847818-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR