3-Fluor-4-iodanilin
Lieferant: ACTIVATE SCIENTIFIC
AS31337G1
AS31337G5
ACTIAS31337G1EA
0
CHF
ACTIAS31337G1
ACTIAS31337G5
3-Fluor-4-iodanilin
3-Fluor-4-iodanilin
Formel:
C₆H₅FIN MW: 237.01 g/mol |
MDL Number:
MFCD07774187 CAS-Nummer: 656-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...