(S)-3-Aminobuttersäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (S)-3-Aminobutyric acid, (S)-3-Aminobutanoic acid

AS35886G1 AS35886G5
ACTIAS35886G1EA 0 CHF
ACTIAS35886G1 ACTIAS35886G5
(S)-3-Aminobuttersäure
(S)-3-Aminobuttersäure
Formel: C₄H₉NO₂
MW: 103,1200 g/mol
MDL Number: MFCD00270347
CAS-Nummer: 3775-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR