(R)-3-Aminobuttersäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (R)-3-Aminobutanoic acid, (R)-3-Aminobutyric acid

AS34208G1 AS34208G5
ACTIAS34208G1EA 0 CHF
ACTIAS34208G1 ACTIAS34208G5
(R)-3-Aminobuttersäure
(R)-3-Aminobuttersäure
Formel: C₄H₉NO₂
MW: 103,1198 g/mol
MDL Number: MFCD00211284
CAS-Nummer: 3775-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR