N-Cbz-3-brom-4-fluoranilin
Lieferant: ACTIVATE SCIENTIFIC
AS23609G1
AS23609G25
AS23609G5
ACTIAS23609G1EA
81
CHF
ACTIAS23609G1
ACTIAS23609G25
ACTIAS23609G5
N-Cbz-3-brom-4-fluoranilin
N-Cbz-3-brom-4-fluoranilin
Formel:
C₁₄H₁₁BrFNO₂ |
MDL Number:
MFCD18071052 CAS-Nummer: 1256633-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...