1,3,5-Trimethyl-1H-pyrazol-4-boronsäure Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

AS15315G1 AS15315G5
ACTIAS15315G1EA 0 CHF
ACTIAS15315G1 ACTIAS15315G5
1,3,5-Trimethyl-1H-pyrazol-4-boronsäure Hydrochlorid
1,3,5-Trimethyl-1H-pyrazol-4-boronsäure Hydrochlorid
Formel: C₆H₁₂BClN₂O₂
MDL Number: MFCD11616524
CAS-Nummer: 1162262-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR