1H-Pyrazol-4-boronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

AS31224G1 AS31224G5
ACTIAS31224G1EA 0 CHF
ACTIAS31224G1 ACTIAS31224G5
1H-Pyrazol-4-boronsäurepinakolester
1H-Pyrazol-4-boronsäurepinakolester
Formel: C₉H₁₅BN₂O₂
MDL Number: MFCD03453063
CAS-Nummer: 269410-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR