1H-Benzimidazol-4-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1H-Benzimidazole-7-carboxylic acid

AS1265G1 AS1265G5
ACTIAS1265G1EA 0 CHF
ACTIAS1265G1 ACTIAS1265G5
1H-Benzimidazol-4-carbonsäure
1H-Benzimidazol-4-carbonsäure
Formel: C₈H₆N₂O₂
MW: 162,14 g/mol
MDL Number: MFCD01823426
CAS-Nummer: 46006-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR