7-Chlorimidazo[4,5-b]pyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 7-Chloro-1H-imidazo[4,5-b]pyridine

AS31942G1 AS31942G5
ACTIAS31942G1EA 0 CHF
ACTIAS31942G1 ACTIAS31942G5
7-Chlorimidazo[4,5-b]pyridin
7-Chlorimidazo[4,5-b]pyridin
Formel: C₆H₄ClN₃
MW: 153.5691 g/mol
MDL Number: MFCD15143399
CAS-Nummer: 6980-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR