Methyl-5-bromimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS51440G1 AS51440G5
ACTIAS51440G1EA 0 CHF
ACTIAS51440G1 ACTIAS51440G5
Methyl-5-bromimidazo[1,2-a]pyridin-2-carboxylat
Methyl-5-bromimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₉H₇BrN₂O₂
MW: 255,681 g/mol
MDL Number: MFCD11109379
CAS-Nummer: 1092351-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR