7-Bromimidazo[1,2-a]pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50986G1 AS50986G5
ACTIAS50986G1EA 0 CHF
ACTIAS50986G1 ACTIAS50986G5
7-Bromimidazo[1,2-a]pyridin
7-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
MDL Number: MFCD07778353
CAS-Nummer: 808744-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR