6-Bromimidazo[1,2-a]pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS2527G1 AS2527G5
ACTIAS2527G1EA 0 CHF
ACTIAS2527G1 ACTIAS2527G5
6-Bromimidazo[1,2-a]pyridin
6-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
MDL Number: MFCD06496200
CAS-Nummer: 6188-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR