3-Bromimidazo[1,2-a]pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS12861G1 AS12861G5
ACTIAS12861G1EA 23 CHF
ACTIAS12861G1 ACTIAS12861G5
3-Bromimidazo[1,2-a]pyridin
3-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
MW: 197,0300 g/mol
MDL Number: MFCD00014556
CAS-Nummer: 4926-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR