6-Chlor-3-methylimidazo[1,2-b]pyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS51742G1 AS51742G5
ACTIAS51742G1EA 0 CHF
ACTIAS51742G1 ACTIAS51742G5
6-Chlor-3-methylimidazo[1,2-b]pyridazin
6-Chlor-3-methylimidazo[1,2-b]pyridazin
Formel: C₇H₆ClN₃
MDL Number: MFCD09991804
CAS-Nummer: 137384-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR