6-Chlor-3-nitroimidazo[1,2-b]pyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS50623G1 AS50623G5
ACTIAS50623G1EA 0 CHF
ACTIAS50623G1 ACTIAS50623G5
6-Chlor-3-nitroimidazo[1,2-b]pyridazin
6-Chlor-3-nitroimidazo[1,2-b]pyridazin
Formel: C₆H₃ClN₄O₂
MW: 198,5700 g/mol
MDL Number: MFCD08276991
CAS-Nummer: 18087-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR