3-Pyrid-4-ylbenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-(Pyridin-4-yl)benzoic acid
AS30230G1
AS30230G5
ACTIAS30230G1EA
0
CHF
ACTIAS30230G1
ACTIAS30230G5
3-Pyrid-4-ylbenzoesäure
3-Pyrid-4-ylbenzoesäure
Formel:
C₁₂H₉NO₂ MW: 199,2054 g/mol |
MDL Number:
MFCD05864808 CAS-Nummer: 4385-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...