3-Pyrid-3-ylbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Pyridin-3-yl-benzoic acid

AS30229G1 AS30229G5
ACTIAS30229G1EA 0 CHF
ACTIAS30229G1 ACTIAS30229G5
3-Pyrid-3-ylbenzoesäure
3-Pyrid-3-ylbenzoesäure
Formel: C₁₂H₉NO₂
MW: 199,2054 g/mol
MDL Number: MFCD03426512
CAS-Nummer: 4385-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR