4-(2-Naphthyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-(Naphthalen-2-yl)phenyl)boronic acid

AS32804G1 AS32804G5
ACTIAS32804G1EA 0 CHF
ACTIAS32804G1 ACTIAS32804G5
4-(2-Naphthyl)phenylboronsäure
4-(2-Naphthyl)phenylboronsäure
Formel: C₁₆H₁₃BO₂
MW: 248.0800 g/mol
MDL Number: MFCD09260454
CAS-Nummer: 918655-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR