4-(1-Naphthyl)phenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-(1-Naphthyl)benzeneboronic acid
AS32803G1
AS32803G5
ACTIAS32803G1EA
0
CHF
ACTIAS32803G1
ACTIAS32803G5
4-(1-Naphthyl)phenylboronsäure
4-(1-Naphthyl)phenylboronsäure
Formel:
C₁₆H₁₃BO₂ MW: 248.09 g/mol |
MDL Number:
MFCD08669639 CAS-Nummer: 870774-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...