3,5-Di-tert-butyl-2-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,5-Di-tert-butylsalicylaldehyde

AS34432G10 AS34432G100
ACTIAS34432G10EA 0 CHF
ACTIAS34432G10 ACTIAS34432G100
3,5-Di-tert-butyl-2-hydroxybenzaldehyd
3,5-Di-tert-butyl-2-hydroxybenzaldehyd
Formel: C₁₅H₂₂O₂
MW: 234,34 g/mol
MDL Number: MFCD00191998
CAS-Nummer: 37942-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR