2-Fluor-6-nitrophenol
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS33408G100EA
0
CHF
ACTIAS33408G100
ACTIAS33408G10
ACTIAS33408G5
2-Fluor-6-nitrophenol
2-Fluor-6-nitrophenol
Formel:
C₆H₄FNO₃ MW: 157,1 g/mol Melting Pt: 91…92 °C |
MDL Number:
MFCD00042446 CAS-Nummer: 1526-17-6 EINECS: 216-199-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...