4-Chlor-3-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS34967G1 AS34967G10 AS34967G5
ACTIAS34967G1EA 0 CHF
ACTIAS34967G1 ACTIAS34967G10 ACTIAS34967G5
4-Chlor-3-nitrobenzaldehyd
4-Chloro-3-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
MW: 185,57 g/mol
Melting Pt: 57…60 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007078
CAS-Nummer: 16588-34-4
EINECS: 240-645-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR