2-Fluor-6-iodbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Fluoro-2-Iodobenzaldehyde

AS30755G1 AS30755G5
ACTIAS30755G1EA 0 CHF
ACTIAS30755G1 ACTIAS30755G5
2-Fluor-6-iodbenzaldehyd
2-Fluoro-6-iodobenzaldehyde
Formel: C₇H₄FIO
MW: 250.01 g/mol
MDL Number: MFCD07371506
CAS-Nummer: 146137-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR