4-Chlor-3-(trifluormethyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Dihydroxy(4-chloro-3-(trifluoromethyl)phenyl)borane, 4-Chloro-3-(trifluoromethyl)benzeneboronic acid, (4-Chloro-3-(trifluoromethyl)phenyl)metaboric acid

AS50136G1 AS50136G5
ACTIAS50136G1EA 0 CHF
ACTIAS50136G1 ACTIAS50136G5
4-Chlor-3-(trifluormethyl)phenylboronsäure
4-Chlor-3-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
MDL Number: MFCD03094999
CAS-Nummer: 176976-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR