4-Chlor-2-(trifluormethyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50121G1 AS50121G5
ACTIAS50121G1EA 0 CHF
ACTIAS50121G1 ACTIAS50121G5
4-Chlor-2-(trifluormethyl)phenylboronsäure
4-Chlor-2-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
MDL Number: MFCD04973086
CAS-Nummer: 313545-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR