L(+)-Arabinose

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS33109G100EA 0 CHF
ACTIAS33109G100
L(+)-Arabinose
L(+)-Arabinose
Formel: C₅H₁₀O₅
MW: 150,13 g/mol
Siedepunkt: 331 °C (1013 hPa)
Melting Pt: 158…160 °C
Dichte: 1,585 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00067709
CAS-Nummer: 5328-37-0
EINECS: 226-214-6
https://ch.vwr.com/stibo/web/std.lang.all/24/52/2302452.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR