2-Chlor-1-fluor-3-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-3-fluoronitrobenzene

AS33742G1 AS33742G10 AS33742G5
ACTIAS33742G1EA 0 CHF
ACTIAS33742G1 ACTIAS33742G10 ACTIAS33742G5
2-Chlor-1-fluor-3-nitrobenzol
2-Chlor-1-fluor-3-nitrobenzol
Formel: C₆H₃ClFNO₂
MW: 175,55 g/mol
MDL Number: MFCD04112549
CAS-Nummer: 21397-07-9
EINECS: 244-365-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR