1-Chlor-2,3-difluor-5-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chloro-4,5-difluoronitrobenzene

AS31509G1 AS31509G5
ACTIAS31509G1EA 0 CHF
ACTIAS31509G1 ACTIAS31509G5
1-Chlor-2,3-difluor-5-nitrobenzol
1-Chloro-2,3-difluoro-5-nitrobenzene
Formel: C₆H₂ClF₂NO₂
MW: 193,54 g/mol
MDL Number: MFCD06658261
CAS-Nummer: 53780-44-2
EINECS: 258-769-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR