1,3-Dichlor-2-fluor-5-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,5-Dichloro-4-fluoro-1-nitrobenzene

AS23104G100 AS23104G25
ACTIAS23104G100EA 0 CHF
ACTIAS23104G100 ACTIAS23104G25
1,3-Dichlor-2-fluor-5-nitrobenzol
1,3-Dichloro-2-fluoro-5-nitrobenzene
Formel: C₆H₂Cl₂FNO₂
MW: 209.99 g/mol
MDL Number: MFCD00129963
CAS-Nummer: 3107-19-5
EINECS: 221-466-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR