3-Fluor-5-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS32675G1 AS32675G5
ACTIAS32675G1EA 0 CHF
ACTIAS32675G1 ACTIAS32675G5
3-Fluor-5-nitrobenzaldehyd
3-Fluoro-5-nitrobenzaldehyde
Formel: C₇H₄FNO₃
MW: 169,1100 g/mol
MDL Number: MFCD11111000
CAS-Nummer: 108159-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR