4-Brom-2-nitrobenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS30594G1
AS30594G5
ACTIAS30594G1EA
0
CHF
ACTIAS30594G1
ACTIAS30594G5
4-Brom-2-nitrobenzaldehyd
4-Bromo-2-nitrobenzaldehyde
Formel:
C₇H₄BrNO₃ MW: 230,0156 g/mol |
MDL Number:
MFCD00463687 CAS-Nummer: 5551-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...