5-Brom-2-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS8458G1 AS8458G5
ACTIAS8458G1EA 0 CHF
ACTIAS8458G1 ACTIAS8458G5
5-Brom-2-nitrobenzaldehyd
5-Brom-2-nitrobenzaldehyd
Formel: C₇H₄BrNO₃
MW: 230,0200 g/mol
MDL Number: MFCD00456508
CAS-Nummer: 20357-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR