2-Chlor-3-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

AS31003G5 AS31003G1
ACTIAS31003G5EA 0 CHF
ACTIAS31003G5 ACTIAS31003G1
2-Chlor-3-nitrophenol
2-Chlor-3-nitrophenol
Formel: C₆H₄ClNO₃
MW: 173,5500 g/mol
MDL Number: MFCD11035882
CAS-Nummer: 603-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR