1,3-Dichlor-2,4-difluor-5-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS35478G1 AS35478G5
ACTIAS35478G1EA 0 CHF
ACTIAS35478G1 ACTIAS35478G5
1,3-Dichlor-2,4-difluor-5-nitrobenzol
1,3-Dichloro-2,4-difluoro-5-nitrobenzene
Formel: C₆HCl₂F₂NO₂
CAS-Nummer: 15952-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR