3-Chlor-5-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS33748G1 AS33748G5
ACTIAS33748G1EA 0 CHF
ACTIAS33748G1 ACTIAS33748G5
3-Chlor-5-nitrobenzaldehyd
3-Chloro-5-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
MW: 185,56 g/mol
MDL Number: MFCD08236780
CAS-Nummer: 22233-54-1
EINECS: 228-830-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR