4-Methoxy-2-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30752G1 AS30752G5
ACTIAS30752G1EA 0 CHF
ACTIAS30752G1 ACTIAS30752G5
4-Methoxy-2-nitrobenzaldehyd
4-Methoxy-2-nitrobenzaldehyde
Formel: C₈H₇NO₄
MW: 181,1455 g/mol
MDL Number: MFCD00024254
CAS-Nummer: 22996-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR