5-Brom-2-chlor-1,3-difluorbenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-3,5-difluorobromobenzene

AS34969G1 AS34969G10 AS34969G5
ACTIAS34969G1EA 0 CHF
ACTIAS34969G1 ACTIAS34969G10 ACTIAS34969G5
5-Brom-2-chlor-1,3-difluorbenzol
5-Brom-2-chlor-1,3-difluorbenzol
Formel: C₆H₂BrClF₂
MW: 227,4300 g/mol
MDL Number: MFCD00143420
CAS-Nummer: 176673-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR