5-Brom-1,3-dichlor-2-fluorbenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,5-Dichloro-4-fluorobromobenzene

AS33722G1 AS33722G5
ACTIAS33722G1EA 0 CHF
ACTIAS33722G1 ACTIAS33722G5
5-Brom-1,3-dichlor-2-fluorbenzol
5-Brom-1,3-dichlor-2-fluorbenzol
Formel: C₆H₂BrCl₂F
MW: 243.8900 g/mol
MDL Number: MFCD09996901
CAS-Nummer: 17318-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR