2-Amino-3-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

AS1975G1 AS1975G5
ACTIAS1975G1EA 0 CHF
ACTIAS1975G1 ACTIAS1975G5
2-Amino-3-methoxypyridin
2-Amino-3-methoxypyridine
Formel: C₆H₈N₂O
MDL Number: MFCD07374874
CAS-Nummer: 10201-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR