2-Fluor-5-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30742G1 AS30742G5
ACTIAS30742G1EA 0 CHF
ACTIAS30742G1 ACTIAS30742G5
2-Fluor-5-hydroxybenzaldehyd
2-Fluoro-5-hydroxybenzaldehyde
Formel: C₇H₅FO₂
MDL Number: MFCD09038469
CAS-Nummer: 103438-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR