4,6-Difluorsalicylaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,4-Difluoro-6-hydroxybenzaldehyde

AS34376G1 AS34376G5
ACTIAS34376G1EA 0 CHF
ACTIAS34376G1 ACTIAS34376G5
4,6-Difluorsalicylaldehyd
4,6-Difluorsalicylaldehyd
Formel: C₇H₄F₂O₂
MW: 158,1023 g/mol
MDL Number: MFCD03701048
CAS-Nummer: 136516-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR